3-(3-Indolyl)propionsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Indol-3-yl)propionsäure, 3-Indolpropionsäure, 3-Indol-3-propionsäure

OS-7428-25G OS-7428-100G
COBBOS-7428-25GEA 38.2 EUR
COBBOS-7428-25G COBBOS-7428-100G
3-(3-Indolyl)propionsäure
3-(3-Indolyl)propionsäure
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,21 g/mol
Schmelzpunkt: 129…132 °C
Dichte: 1,356 g/cm³ (20 °C)
MDL: MFCD00005660
CAS-Nummer: 830-96-6
EINECS: 212-600-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR