(S)-(-)-3-Butin-2-ol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-(-)-3-Hydroxy-1-butin, (S)-(-)-But-3-in-2-ol
M-Clarity™ Qualitätsstufe = MQ100
464007-1G
464007-5G
SIAL464007-1GEA
94.2
EUR
SIAL464007-1G
SIAL464007-5G
(S)-(-)-3-Butin-2-ol, Sigma-Aldrich®
(S)-(-)-3-Butin-2-ol
Formel:
C₄H₆O Molecular Weight: 70,09 g/mol |
MDL:
MFCD00190166 CAS-Nummer: 2914-69-4 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...