1-Chlor-3,5-dibrombenzol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene
OT-0552-25G
OT-0552-5G
OT-0552-100G
COBBOT-0552-25GEA
102
EUR
COBBOT-0552-25G
COBBOT-0552-5G
COBBOT-0552-100G
1-Chlor-3,5-dibrombenzol
1-Chloro-3,5-dibromobenzene
Formel:
C₆H₃Br₂Cl Molecular Weight: 270.35 g/mol |
MDL:
MFCD00070765 CAS-Nummer: 14862-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...