4-Brom-2-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

722421-1G 722421-5G
SIAL722421-1GEA 26.9 EUR
SIAL722421-1G SIAL722421-5G
4-Brom-2-fluorpyridin, Sigma-Aldrich®
4-Bromo-2-fluoropyridine
Formel: C₅H₃BrFN
Molecular Weight: 175,99 g/mol
MDL: MFCD04112504
CAS-Nummer: 128071-98-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR