2-Chlor-4-(trifluormethoxy)anilin Hydrochlorid
Lieferant: COMBI-BLOCKS
OT-0898-25G
OT-0898-5G
COBBOT-0898-25GEA
111
EUR
COBBOT-0898-25G
COBBOT-0898-5G
2-Chlor-4-(trifluormethoxy)anilin Hydrochlorid
2-Chlor-4-(trifluormethoxy)anilin Hydrochlorid
Formel:
C₇H₆Cl₂F₃NO |
MDL:
MFCD12546529 CAS-Nummer: 1187386-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...