4-Brom-1-naphthylamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Amino-4-bromnaphthalin
M-Clarity™ Qualitätsstufe = MQ100
A43405-25G
A43405-5G
SIALA43405-25GEA
333
EUR
SIALA43405-25G
SIALA43405-5G
4-Brom-1-naphthylamin, Sigma-Aldrich®
4-Brom-1-naphthylamin
Formel:
C₁₀H₈BrN Molecular Weight: 222,08 g/mol |
MDL:
MFCD00004023 CAS-Nummer: 2298-07-9 EINECS: 218-944-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...