N-Methyl-2-(trifluormethoxy)anilin
Lieferant: COMBI-BLOCKS
HC-2921-25G
HC-2921-5G
HC-2921-1G
COBBHC-2921-25GEA
451
EUR
COBBHC-2921-25G
COBBHC-2921-5G
COBBHC-2921-1G
N-Methyl-2-(trifluormethoxy)anilin
N-Methyl-2-(trifluormethoxy)anilin
Formel:
C₈H₈F₃NO Molecular Weight: 191.15 g/mol |
MDL:
MFCD00052334 CAS-Nummer: 175278-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...