N-Methyl-4-(trifluormethoxy)anilin
Lieferant: COMBI-BLOCKS
HC-2920-5G
HC-2920-1G
COBBHC-2920-5GEA
127
EUR
COBBHC-2920-5G
COBBHC-2920-1G
N-Methyl-4-(trifluormethoxy)anilin
N-Methyl-4-(trifluormethoxy)anilin
Formel:
C₈H₈F₃NO Molecular Weight: 191,15 g/mol |
MDL:
MFCD00052332 CAS-Nummer: 41419-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...