1-Butyl-3-methylimidazoliumiodid, Sigma-Aldrich®

Lieferant: Merck

713066-50G 713066-5G
SIAL713066-50GEA 343 EUR
SIAL713066-50G SIAL713066-5G
1-Butyl-3-methylimidazoliumiodid, Sigma-Aldrich®
1-Butyl-3-methylimidazolium iodide
Formel: C₈H₁₅IN₂
Molecular Weight: 266,12 g/mol
Dichte: 1,49 g/cm³ (20 °C)
Flammpunkt: 223 °C
MDL: MFCD08276307
CAS-Nummer: 65039-05-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR