2-Amino-6-chlorbenzothiazol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chlorobenzothiazol-2-ylamine, 6-Chloro-1,3-benzothiazol-2-amine

HC-2013-25G HC-2013-100G
COBBHC-2013-25GEA 37.5 EUR
COBBHC-2013-25G COBBHC-2013-100G
2-Amino-6-chlorbenzothiazol
2-Amino-6-chlorobenzothiazole
Formel: C₇H₅ClN₂S
Molecular Weight: 184,65 g/mol
Siedepunkt: 344 °C (1013 hPa)
Schmelzpunkt: 160…163 °C
MDL: MFCD00053557
CAS-Nummer: 95-24-9
EINECS: 202-402-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR