3-(o-Tolyl)propionsäure 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(2-Methylphenyl)propansäure, 3-(2-Methylphenyl)propionsäure
H34375.03
H34375.09
H34375.03EA
32
EUR
H34375.03
H34375.09
3-(o-Tolyl)propionsäure 96%
3-(o-Tolyl)propionsäure
Purity: 96%
CAS No.: 22084-89-5
Molecular Formula: C10H12O2
Formula Weight: 164.20
Melting Point: 101-103°
MDL No.: MFCD00079773
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079773 CAS-Nummer: 22084-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...