3-(m-Tolyl)propionsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(3-Methylphenyl)propionsäure, 3-(3-Methylphenyl)propansäure

H34395.03 H34395.06
H34395.03EA 57.3 EUR
H34395.03 H34395.06
3-(m-Tolyl)propionsäure 96%
3-(m-Tolyl)propionsäure

Purity: 96% CAS No.: 3751-48-2 Molecular Formula: C10H12O2 Formula Weight: 164.20 Melting Point: 42-44° MDL No.: MFCD00060326

Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2 g/mol
Schmelzpunkt: 42…44 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060326
CAS-Nummer: 3751-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR