6-Indolcarbaldehyd
Lieferant: COMBI-BLOCKS
Synonyme:
Indol-6-carboxaldehyd
IN-0017-5G
IN-0017-25G
IN-0017-1G
COBBIN-0017-5GEA
97.2
EUR
COBBIN-0017-5G
COBBIN-0017-25G
COBBIN-0017-1G
6-Indolcarbaldehyd
6-Indolcarbaldehyd
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol |
MDL:
MFCD02179596 CAS-Nummer: 1196-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...