5-Chlor-2-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(5-Chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-Chloro-2-fluorobenzeneboronic acid pinacol ester

PN-2388-5G PN-2388-1G
COBBPN-2388-5GEA 346 EUR
COBBPN-2388-5G COBBPN-2388-1G
5-Chlor-2-fluorphenylboronsäurepinakolester
5-Chlor-2-fluorphenylboronsäurepinakolester
Formel: C₁₂H₁₅BClFO₂
Molecular Weight: 256,5087 g/mol
MDL: MFCD16036133
CAS-Nummer: 1190129-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR