2-Bromimidazol 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Brom-1H-imidazol
H52553.03
H52553.06
H52553.03EA
201
EUR
H52553.03
H52553.06
2-Bromimidazol 95%
2-Bromoimidazole
Formel:
C₃H₃BrN₂ Molecular Weight: 146,97 g/mol Schmelzpunkt: 197 °C |
MDL:
MFCD02179526 CAS-Nummer: 16681-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...