3,4,5-Trihydroxybenzaldehyd Monohydrat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3,4,5-Trihydroxybenzaldehyde hydrate
M-Clarity™ Qualitätsstufe = MQ100
259594-1G
259594-5G
SIAL259594-1GEA
112
EUR
SIAL259594-1G
SIAL259594-5G
3,4,5-Trihydroxybenzaldehyd Monohydrat, Sigma-Aldrich®
3,4,5-Trihydroxybenzaldehyd Monohydrat
Formel:
C₇H₆O₄·1H₂O Molecular Weight: 172,14 g/mol |
MDL:
MFCD00149124 CAS-Nummer: 207742-88-9 EINECS: 237-168-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...