2-Chlor-5-(trifluormethyl)benzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-5-(trifluormethyl)benzol-1-sulfonylchlorid, 2-Chlor-5-(trifluormethyl)benzolsulfochlorid, 2-Chlor-5-(trifluormethyl)benzolsulfonsäurechlorid

004537-1G 004537-5G 004537-25G
FLUO004537-1GEA 0 EUR
FLUO004537-1G FLUO004537-5G FLUO004537-25G
2-Chlor-5-(trifluormethyl)benzolsulfonylchlorid
2-Chlor-5-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃Cl₂F₃O₂S
Molecular Weight: 279.07
MDL: MFCD00052310
CAS-Nummer: 54090-08-3
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR