3-Acetylbenzolsulfonylchlorid 97
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Acetylbenzol-1-sulfonylchlorid, 3-Acetylbenzolsulfochlorid, 3-Acetylbenzolsulfonsäurechlorid
H32447.03
H32447.06
H32447.03EA
71.7
EUR
H32447.03
H32447.06
3-Acetylbenzolsulfonylchlorid 97
3-Acetylbenzolsulfonylchlorid
Purity: 97%
CAS No.: 73035-16-2
Molecular Formula: C8H7ClO3S
Formula Weight: 218.66
Melting Point: 42-43°
UN No.: 3261
MDL No.: MFCD03424979
Formel:
C₈H₇ClO₃S Molecular Weight: 218.66 Schmelzpunkt: 42…43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424979 CAS-Nummer: 73035-16-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...