2-Fluor-6-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

BB-5722-1G BB-5722-250M
COBBBB-5722-1GEA 0 EUR
COBBBB-5722-1G COBBBB-5722-250M
2-Fluor-6-(trifluormethyl)pyridin
2-Fluoro-6-(trifluoromethyl)pyridine
Formel: C₆H₃F₄N
MDL: MFCD03092901
CAS-Nummer: 94239-04-0
EINECS: 428-100-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR