3'-Fluor-4'-methylacetophenon

Lieferant: FLUOROCHEM

005782-1G 005782-5G
FLUO005782-1GEA 45.9 EUR
FLUO005782-1G FLUO005782-5G
3'-Fluor-4'-methylacetophenon
3'-Fluor-4'-methylacetophenon
Formel: C₉H₉FO
MDL: MFCD01631411
CAS-Nummer: 42444-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR