3'-Trifluormethoxyacetophenon

Lieferant: Thermo Scientific

Synonyme: 1-[3-(Trifluormethoxy)phenyl]-1-ethanon

SB01485.1 SB01485.10
MAYBSB01485.1EA 0 EUR
MAYBSB01485.1 MAYBSB01485.10
3'-Trifluormethoxyacetophenon
3'-Trifluormethoxyacetophenon
Formel: C₉H₇F₃O₂
MDL: MFCD00042403
CAS-Nummer: 170141-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR