D-α-Tocopherol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E Alkohol, (+)-α-Tocopherol, D(+)-α-Tocopherol, Vitamin E

M-Clarity™ Qualitätsstufe = MQ200

T1539-100G T1539-25G
SIALT1539-100GEA 359 EUR
SIALT1539-100G SIALT1539-25G
D-α-Tocopherol, Sigma-Aldrich®
D-α-Tocopherol
Formel: C₂₉H₅₀O₂
Molecular Weight: 430,71 g/mol
MDL: MFCD00072045
CAS-Nummer: 59-02-9
EINECS: 233-466-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR