Zink(II)acetylacetonat Hydrat ≥98%
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
Synonyme:
Di(2,4-pentandionato-O,O')zink(II) Hydrat, Bis(2,4-pentandionato-O,O')zink(II) Hydrat, Zink(II)-2,4-pentandionat Hydrat, Zink(II)-bis(2,4-pentandionato-O,O') Hydrat
93-3031-50G
93-3031-250G
STRE93-3031-50GEA
58.8
EUR
STRE93-3031-50G
STRE93-3031-250G
Zink(II)acetylacetonat Hydrat ≥98%
Zink(II)acetylacetonat Hydrat
Formel:
C₁₀H₁₄O₄Zn·H₂O Molecular Weight: 281,62 g/mol |
MDL:
MFCD00149060 CAS-Nummer: 108503-47-5 EINECS: 237-860-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...