Ethyl-4-brombutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 4-bromobutyrate

M-Clarity™ Qualitätsstufe = MQ200

167118-250G 167118-50G
SIAL167118-250GEA 196 EUR
SIAL167118-250G SIAL167118-50G
Ethyl-4-brombutyrat, Sigma-Aldrich®
Ethyl-4-brombutyrat
Formel: C₆H₁₁BrO₂
Molecular Weight: 195,06 g/mol
MDL: MFCD00000259
CAS-Nummer: 2969-81-5
EINECS: 221-005-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR