5-tert-Butylsalicylaldehyd

Lieferant: FLUOROCHEM

Synonyme: 5-(tert-Butyl)-2-hydroxybenzaldehyde, 5-tert-Butyl-2-hydroxybenzaldehyde

031709-10G 031709-20G 031709-50G 031709-5G
FLUO031709-10GEA 554 EUR
FLUO031709-10G FLUO031709-20G FLUO031709-50G FLUO031709-5G
5-tert-Butylsalicylaldehyd
5-tert-Butylsalicylaldehyd
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,23 g/mol
MDL: MFCD01321335
CAS-Nummer: 2725-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR