1-Methyl-2-piperidon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Methylpiperidin-2-one, N-Methyl-2-piperidone
M-Clarity™ Qualitätsstufe = MQ100
M73788-10G
M73788-25G
SIALM73788-10GEA
221
EUR
SIALM73788-10G
SIALM73788-25G
1-Methyl-2-piperidon, Sigma-Aldrich®
1-Methyl-2-piperidon
Formel:
C₆H₁₁NO Molecular Weight: 113,16 g/mol |
MDL:
MFCD00006555 CAS-Nummer: 931-20-4 EINECS: 213-231-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...