1-Methyl-4-piperidon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Methyl-4-piperidone
M-Clarity™ Qualitätsstufe = MQ100
130036-100ML
130036-500ML
130036-5ML
SIAL130036-100MLEA
122
EUR
SIAL130036-100ML
SIAL130036-500ML
SIAL130036-5ML
1-Methyl-4-piperidon, Sigma-Aldrich®
1-Methyl-4-piperidon
Formel:
C₆H₁₁NO Molecular Weight: 113,16 g/mol |
MDL:
MFCD00006191 CAS-Nummer: 1445-73-4 EINECS: 215-895-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...