Dinatriummalonat ≥99%
Lieferant: Thermo Scientific
Synonyme:
Sodium malonate dibasic, Malonic acid disodium salt
44841.14
44841.22
44841.14EA
38
EUR
44841.14
44841.22
Dinatriummalonat ≥99%
Dinatriummalonat
Purity: 99%
CAS No.: 141-95-7
Molecular Formula: CH2(COONa)2
Formula Weight: 148.03
Physical Form: Powder
Formel:
C₃H₂Na₂O₄ Molecular Weight: 148,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002708 CAS-Nummer: 141-95-7 EINECS: 205-514-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...