Dinatriummalonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Sodium malonate dibasic, Malonic acid disodium salt
63409-100G
63409-500G
SIAL63409-100GEA
113
EUR
SIAL63409-100G
SIAL63409-500G
Dinatriummalonat, Sigma-Aldrich®
Dinatriummalonat
Formel:
C₃H₂Na₂O₄ Molecular Weight: 148,03 g/mol |
MDL:
MFCD00002708 CAS-Nummer: 141-95-7 EINECS: 205-514-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...