2,6-Dibrom-4-tert-butylanilin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(tert-Butyl)-2,6-dibromoaniline, 2,6-Dibromo-4-(tert-butyl)aniline

AN-1242-25G AN-1242-5G AN-1242-1G
COBBAN-1242-25GEA 262 EUR
COBBAN-1242-25G COBBAN-1242-5G COBBAN-1242-1G
2,6-Dibrom-4-tert-butylanilin
2,6-Dibrom-4-tert-butylanilin
Formel: C₁₀H₁₃Br₂N
Molecular Weight: 307.03 g/mol
MDL: MFCD06657952
CAS-Nummer: 10546-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR