4-Brom-3,5-dimethylanisol

Lieferant: FLUOROCHEM

045085-5G 045085-25G
FLUO045085-5GEA 57.9 EUR
FLUO045085-5G FLUO045085-25G
4-Brom-3,5-dimethylanisol
4-Brom-3,5-dimethylanisol
Formel: C₉H₁₁BrO
MDL: MFCD00461692
CAS-Nummer: 6267-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR