4-Chlorbutyrylchlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Chlorobutanoyl Chloride
A12249.09
A12249.0B
A12249.30
A12249.09EA
27.4
EUR
A12249.09
A12249.0B
A12249.30
4-Chlorbutyrylchlorid ≥98%
4-Chlorbutyrylchlorid
Purity: 98%
CAS No.: 4635-59-0
Molecular Formula: C4H6Cl2O
Formula Weight: 141.00
Flash Point: 85°(185°F)
Refractive Index: 1.4610
Odour: Acrid
Melting Point: -49°
Boiling Point: 173-174°
Beilstein Reference: 773860
UN No.: 2927
MDL No.: MFCD00000754
Formel:
C₄H₆Cl₂O Molecular Weight: 141 g/mol Siedepunkt: 173…174 °C Schmelzpunkt: –49 °C Dichte: 1,258 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000754 CAS-Nummer: 4635-59-0 EINECS: 225-059-1 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...