2-Isopropylphenylisocyanat ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Isopropylphenylisocyanate
L10618.03
L10618.06
L10618.14
L10618.03EA
26.2
EUR
L10618.03
L10618.06
L10618.14
2-Isopropylphenylisocyanat ≥97%
2-Isopropylphenylisocyanat
Purity: 97%
CAS No.: 56309-56-9
Molecular Formula: C10H11NO
Formula Weight: 161.21
Storage Temperature: 2°C to 8°C
Flash Point: 87°(188°F)
Refractive Index: 1.5230
Boiling Point: 66-67°/5mm
Beilstein Reference: 3588928
UN No.: 2206
MDL No.: MFCD00013850
Formel:
C₁₀H₁₁NO Molecular Weight: 161,2 g/mol Siedepunkt: 66…67 °C (5 mmHg) Dichte: 1,019 Flammpunkt: 87 °C (188 °F) |
MDL:
MFCD00013850 CAS-Nummer: 56309-56-9 EINECS: 260-105-4 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...