2-Ethyl-6-methylphenylisocyanat
Lieferant: Thermo Scientific
Synonyme:
2-Ethyl-6-methylphenylisocyanate
SB01845.1
SB01845.50
MAYBSB01845.1EA
0
EUR
MAYBSB01845.1
MAYBSB01845.50
2-Ethyl-6-methylphenylisocyanat
2-Ethyl-6-methylphenylisocyanat
Formel:
C₁₀H₁₁NO Molecular Weight: 161,2 g/mol |
MDL:
MFCD00013855 CAS-Nummer: 75746-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...