DL-Threonin ≥99%

Lieferant: Thermo Fisher Scientific

A10606.14 A10606.22 A10606.36
A10606.14EA 41.6 EUR
A10606.14 A10606.22 A10606.36
DL-Threonin ≥99%
DL-Threonin

Purity: 99% CAS No.: 80-68-2 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: ca 244° dec. Beilstein Reference: 1721647 Merck Reference: 14,9380 MDL No.: MFCD00063722

Formel: C₄H₉NO₃
Molecular Weight: 119.12 g/mol
Schmelzpunkt: ∼ 244 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00063722
CAS-Nummer: 80-68-2
EINECS: 201-300-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR