D-allo-Threonin 99

Lieferant: Thermo Fisher Scientific

Synonyme: H-D-Allo-Thr-OH, (2R,3R)-2-Amino-3-hydroxybutanoic acid

H27050.MD H27050.03
H27050.MDEA 126 EUR
H27050.MD H27050.03
D-allo-Threonin 99
D-allo-Threonin

Purity: 99% CAS No.: 24830-94-2 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: 268° dec. Beilstein Reference: 1721644

Formel: C₄H₉NO₃
Molecular Weight: 119.12 g/mol
Schmelzpunkt: 268 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004526
CAS-Nummer: 24830-94-2
EINECS: 246-488-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR