D-Threonin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
H-D-Thr-OH, (2R,3S)-2-Amino-3-hydroxybutanoic acid
M-Clarity™ Qualitätsstufe = MQ200
T8250-100G
T8250-25G
T8250-5G
SIALT8250-100GEA
912
EUR
SIALT8250-100G
SIALT8250-25G
SIALT8250-5G
D-Threonin, Sigma-Aldrich®
D-Threonin
Formel:
C₄H₉NO₃ Molecular Weight: 119,12 g/mol |
MDL:
MFCD00064269 CAS-Nummer: 632-20-2 EINECS: 211-171-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...