D-Threonin

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-D-Thr-OH, (2R,3S)-2-Amino-3-hydroxybutanoic acid

4031299.0025 4031299.0100
BACE4031299.0025EA 0 EUR
BACE4031299.0025 BACE4031299.0100
D-Threonin
D-Threonin
Formel: C₄H₉NO₃
Molecular Weight: 119.12 g/mol
MDL: MFCD00064269
CAS-Nummer: 632-20-2
EINECS: 211-171-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR