D-Threonin ≥99%

Lieferant: Thermo Scientific

Synonyme: H-D-Thr-OH, (2R,3S)-2-Amino-3-hydroxybutanoic acid

B21177.06 B21177.14 B21177.22
B21177.06EA 40.1 EUR
B21177.06 B21177.14 B21177.22
D-Threonin ≥99%
D-Threonin

Purity: 99% CAS No.: 632-20-2 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: ca 274° dec. Beilstein Reference: 1721643

Formel: C₄H₉NO₃
Molecular Weight: 119,12 g/mol
Schmelzpunkt: ∼ 274 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064269
CAS-Nummer: 632-20-2
EINECS: 211-171-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR