L-Threonin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

M-Clarity™ Qualitätsstufe = MQ200

T8625-100G T8625-10G T8625-1G T8625-1KG T8625-25G T8625-500G T8625-5KG
SIALT8625-100GEA 175 EUR
SIALT8625-100G SIALT8625-10G SIALT8625-1G SIALT8625-1KG SIALT8625-25G SIALT8625-500G SIALT8625-5KG
L-Threonin, Sigma-Aldrich®
L-Threonin
Formel: C₄H₉NO₃
Molecular Weight: 119,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064270
CAS-Nummer: 72-19-5
EINECS: 200-774-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR