L-allo-Threonin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
H-allo-Thr-OH, L(+)-allo-Threonine, Allo-Thr-OH
4008379.0250
4008379.1000
BACE4008379.0250EA
0
EUR
BACE4008379.0250
BACE4008379.1000
L-allo-Threonin
L-allo-Threonin
Formel:
C₄H₉NO₃ Molecular Weight: 119.12 g/mol |
MDL:
MFCD00064268 CAS-Nummer: 28954-12-3 EINECS: 249-327-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...