6-Methyl-1,2,3,4-tetrahydrochinolin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,2,3,4-Tetrahydro-6-methylquinoline

A15245.09 A15245.18
A15245.09EA 146 EUR
A15245.09 A15245.18
6-Methyl-1,2,3,4-tetrahydrochinolin ≥98%
6-Methyl-1,2,3,4-tetrahydrochinolin

Purity: 98% CAS No.: 91-61-2 Molecular Formula: C10H13N Formula Weight: 147.22 Melting Point: 33-37° Boiling Point: 262-263°/712mm MDL No.: MFCD00023887

Formel: C₁₀H₁₃N
Molecular Weight: 147,22 g/mol
Siedepunkt: 262…263 °C (712 mmHg)
Schmelzpunkt: 33…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023887
CAS-Nummer: 91-61-2
EINECS: 202-083-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR