Isobutyrylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

139122-5G 139122-100G 139122-500G
SIAL139122-5GEA 39 EUR
SIAL139122-5G SIAL139122-100G SIAL139122-500G
Isobutyrylchlorid, Sigma-Aldrich®
Isobutyrylchlorid
Formel: C₄H₇ClO
Molecular Weight: 106.55 g/mol
MDL: MFCD00000717
CAS-Nummer: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR