Isobutylisobutyrat ≥98%

Lieferant: Thermo Scientific
Achtung

L04655.AK L04655.AU
L04655.AKEA 53 EUR
L04655.AK L04655.AU
Isobutylisobutyrat ≥98%
Isobutylisobutyrat

Purity: 98% CAS No.: 97-85-8 Molecular Formula: (CH3)2CHCOOCH2CH(CH3)2 Formula Weight: 144.22 Flash Point: 37°(98°F) Refractive Index: 1.3995 Odour: Fruit-like Melting Point: -81° Boiling Point: 147-149° Beilstein Reference: 1701355 Merck Reference: 14,5145 UN No.: 2528 MDL No.: MFCD00008916

Formel: C₈H₁₆O₂
Molecular Weight: 144,21 g/mol
Siedepunkt: 147…149 °C
Schmelzpunkt: –81 °C
Dichte: 0,855
Flammpunkt: 37 °C (98 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008916
CAS-Nummer: 97-85-8
EINECS: 202-612-5
UN: 2528
ADR: 3,III
Merck Index: 14,5145

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR