6-Chlor-2-fluor-m-toluylsäure
Lieferant: FLUOROCHEM
Synonyme:
6-Chloro-2-fluoro-3-methylbenzoic acid
005647-5G
005647-10G
FLUO005647-5GEA
219
EUR
FLUO005647-5G
FLUO005647-10G
6-Chlor-2-fluor-m-toluylsäure
6-Chlor-2-fluor-m-toluylsäure
Formel:
C₈H₆ClFO₂ Molecular Weight: 188,59 g/mol |
MDL:
MFCD01631369 CAS-Nummer: 32890-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...