6-Chlor-2-fluor-m-kresol

Lieferant: FLUOROCHEM

Synonyme: 6-Chloro-2-fluoro-3-methylphenol

005659-5G 005659-1G
FLUO005659-5GEA 272 EUR
FLUO005659-5G FLUO005659-1G
6-Chlor-2-fluor-m-kresol
6-Chlor-2-fluor-m-kresol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD01631376
CAS-Nummer: 261762-91-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR