2-Chlor-6-fluor-m-kresol

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-6-fluoro-3-methylphenol, 2-Choro-6-fluoro-3-methylphenol

005658-5G 005658-1G
FLUO005658-5GEA 0 EUR
FLUO005658-5G FLUO005658-1G
2-Chlor-6-fluor-m-kresol
2-Chlor-6-fluor-m-kresol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD01631366
CAS-Nummer: 261762-90-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR