2',4'-Dimethoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4952-1G CA-4952-250M
COBBCA-4952-1GEA 742 EUR
COBBCA-4952-1G COBBCA-4952-250M
2',4'-Dimethoxybiphenyl-3-carbonsäure
2',4'-Dimethoxybiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₄
MDL: MFCD04117424
CAS-Nummer: 728919-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR