2',5'-Dimethoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4935-5G CA-4935-1G
COBBCA-4935-5GEA 884 EUR
COBBCA-4935-5G COBBCA-4935-1G
2',5'-Dimethoxybiphenyl-3-carbonsäure
2',5'-Dimethoxybiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₄
MDL: MFCD11932236
CAS-Nummer: 925909-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR