3',4'-Dimethoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3',4'-Dimethoxy-[1,1'-biphenyl]-3-carboxylic acid

CA-4933-5G CA-4933-1G CA-4933-250M
COBBCA-4933-5GEA 1180 EUR
COBBCA-4933-5G COBBCA-4933-1G COBBCA-4933-250M
3',4'-Dimethoxybiphenyl-3-carbonsäure
3',4'-Dimethoxybiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₄
Molecular Weight: 258,27 g/mol
MDL: MFCD03424611
CAS-Nummer: 676348-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR